2-Amino-4,6-dichlorpyrimidin 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,6-Dichloropyrimidin-2-ylamine

OR11290-100G OR11290-25G OR11290-500G
APOSOR11290-100GEA 24.8 EUR
APOSOR11290-100G APOSOR11290-25G APOSOR11290-500G
2-Amino-4,6-dichlorpyrimidin 95%
2-Amino-4,6-dichloropyrimidine
Formel: C₄H₃Cl₂N₃
Molecular Weight: 163,99 g/mol
Schmelzpunkt: 218…222 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006090
CAS-Nummer: 56-05-3
EINECS: 200-253-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR