2,3,5-Triiodbenzoesäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: TIBA

OR11299-100G OR11299-25G
APOSOR11299-100GEA 272 EUR
APOSOR11299-100G APOSOR11299-25G
2,3,5-Triiodbenzoesäure 98%
2,3,5-Triiodbenzoesäure
Formel: C₇H₃I₃O₂
Molecular Weight: 499,81 g/mol
Schmelzpunkt: 220…229 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002420
CAS-Nummer: 88-82-4
EINECS: 201-859-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR