5-Brom-2-hydroxybenzophenon
Lieferant: Apollo Scientific
|
Achtung
|
OR1131-5G
OR1131-25G
APOSOR1131-5GEA
230
EUR
APOSOR1131-5G
APOSOR1131-25G
5-Brom-2-hydroxybenzophenon
5-Brom-2-hydroxybenzophenon
Formel:
C₁₃H₉BrO₂ Molecular Weight: 277,11 g/mol Schmelzpunkt: 108…111 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00525062 CAS-Nummer: 55082-33-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...