2-Amino-6-brombenzothiazol 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR11314-100G
APOSOR11314-100GEA
162
EUR
APOSOR11314-100G
2-Amino-6-brombenzothiazol 98%
2-Amino-6-bromobenzothiazole
Formel:
C₇H₅BrN₂S Molecular Weight: 229,1 g/mol Schmelzpunkt: 213…217 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00152229 CAS-Nummer: 15864-32-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...