2H-1,2,4-Triazole-3-carbonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 2H-1,2,4-triazole-3-carbonitrile

OR11327-5G OR11327-25G
APOSOR11327-5GEA 21.9 EUR
APOSOR11327-5G APOSOR11327-25G
2H-1,2,4-Triazole-3-carbonitrile
2H-1,2,4-Triazole-3-carbonitrile
Formel: C₃H₂N₄
Molecular Weight: 94.0700 g/mol
Schmelzpunkt: 187…190 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11505654
CAS-Nummer: 3641-10-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR