4-(2-Chlorphenoxy)anilin

Lieferant: Apollo Scientific
Achtung

OR11348-25G OR11348-5G
APOSOR11348-25GEA 299 EUR
APOSOR11348-25G APOSOR11348-5G
4-(2-Chlorphenoxy)anilin
4-(2-Chlorphenoxy)anilin
Formel: C₁₂H₁₀ClNO
Molecular Weight: 219,67 g/mol
Schmelzpunkt: 74…78 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01733272
CAS-Nummer: 56705-85-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR