4-(2-Methylphenoxy)anilin

Lieferant: Apollo Scientific
Achtung

OR11350-25G OR11350-5G
APOSOR11350-25GEA 174 EUR
APOSOR11350-25G APOSOR11350-5G
4-(2-Methylphenoxy)anilin
4-(2-Methylphenoxy)anilin
Formel: C₁₃H₁₃NO
Molecular Weight: 199.25 g/mol
Schmelzpunkt: 61…62 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02663364
CAS-Nummer: 56705-83-0
EINECS: 260-349-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR