3-Brom-5-chlorbenzonitril
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-Bromo-5-chlorobenzonitile
OR11380-10G
OR11380-1G
OR11380-5G
APOSOR11380-10GEA
40.6
EUR
APOSOR11380-10G
APOSOR11380-1G
APOSOR11380-5G
3-Brom-5-chlorbenzonitril
3-Brom-5-chlorbenzonitril
Formel:
C₇H₃BrClN Molecular Weight: 216,46 g/mol Schmelzpunkt: 70,7…71.1 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06200840 CAS-Nummer: 304854-55-5 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...