3,5-Dibromphenol 98%
Lieferant: Apollo Scientific
|
Gefahr
|
OR11480-100G
OR11480-25G
OR11480-5G
APOSOR11480-100GEA
546
EUR
APOSOR11480-100G
APOSOR11480-25G
APOSOR11480-5G
3,5-Dibromphenol 98%
3,5-Dibromphenol
Formel:
C₆H₄Br₂O Molecular Weight: 251,91 g/mol Schmelzpunkt: 79…83 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06411376 CAS-Nummer: 626-41-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...