7-Ethoxycumarin
Lieferant: Apollo Scientific
Synonyme:
7-Ethoxy-2H-chromen-2-one
OR1150-100MG
OR1150-1G
OR1150-250MG
APOSOR1150-100MGEA
21.9
EUR
APOSOR1150-100MG
APOSOR1150-1G
APOSOR1150-250MG
7-Ethoxycumarin
7-Ethoxycumarin
Formel:
C₁₁H₁₀O₃ Molecular Weight: 190,20 g/mol Schmelzpunkt: 88…91 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006877 CAS-Nummer: 31005-02-4 EINECS: 250-429-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...