5-Chlor-2-nitroanisol 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR11523-25G
OR11523-5G
APOSOR11523-25GEA
77
EUR
APOSOR11523-25G
APOSOR11523-5G
5-Chlor-2-nitroanisol 98%
5-Chlor-2-nitroanisol
Formel:
C₇H₆ClNO₃ Molecular Weight: 187,58 g/mol Schmelzpunkt: 70…72 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007288 CAS-Nummer: 6627-53-8 EINECS: 229-594-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...