5-Amino-2-brombenzonitril 98%

Lieferant: Apollo Scientific
Achtung

OR11524-1G OR11524-25G OR11524-5G
APOSOR11524-1GEA 21.9 EUR
APOSOR11524-1G APOSOR11524-25G APOSOR11524-5G
5-Amino-2-brombenzonitril 98%
5-Amino-2-brombenzonitril
Formel: C₇H₅BrN₂
Molecular Weight: 197,0320 g/mol
Schmelzpunkt: 96…97 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00234259
CAS-Nummer: 72115-09-4
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR