2-(N-tert-Butylamino)-3,6,7-trichlorchinoxalin 96%
Lieferant: Apollo Scientific
OR11539-1G
OR11539-250MG
APOSOR11539-1GEA
285
EUR
APOSOR11539-1G
APOSOR11539-250MG
2-(N-tert-Butylamino)-3,6,7-trichlorchinoxalin 96%
2-(N-tert-Butylamino)-3,6,7-trichlorchinoxalin
Formel:
C₁₂H₁₂Cl₃N₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09475905 CAS-Nummer: 281211-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...