2-Acetylamino-5-brom-6-methylpyridin 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Acetamido-5-bromo-6-methylpyridine, 6-Acetamido-3-bromo-2-methylpyridine
OR11546-1G
OR11546-5G
OR11546-25G
APOSOR11546-1GEA
21.9
EUR
APOSOR11546-1G
APOSOR11546-5G
APOSOR11546-25G
2-Acetylamino-5-brom-6-methylpyridin 98%
2-Acetylamino-5-brom-6-methylpyridin
Formel:
C₈H₉BrN₂O Molecular Weight: 229.0700 g/mol Schmelzpunkt: 154…158 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03909755 CAS-Nummer: 142404-84-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...