4-Brom-3-chlor-2-methylanilin 96%
Lieferant: Apollo Scientific
|
Achtung
|
OR11564-5G
OR11564-100G
OR11564-25G
APOSOR11564-5GEA
68.3
EUR
APOSOR11564-5G
APOSOR11564-100G
APOSOR11564-25G
4-Brom-3-chlor-2-methylanilin 96%
4-Brom-3-chlor-2-methylanilin
Formel:
C₇H₇BrClN Molecular Weight: 220,4942 g/mol Schmelzpunkt: 55 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07369999 CAS-Nummer: 627531-47-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...