Methyl-5-bromo-1H-pyrrole-2-carboxylate

Lieferant: Apollo Scientific

Synonyme: Methyl 5-bromo-1H-pyrrole-2-carboxylate

OR11588-1G
APOSOR11588-1GEA 62.5 EUR
APOSOR11588-1G
Methyl-5-bromo-1H-pyrrole-2-carboxylate
Methyl-5-bromo-1H-pyrrole-2-carboxylate
Formel: C₆H₆BrNO₂
Molecular Weight: 204,0213 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09801030
CAS-Nummer: 934-07-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR