4-Brom-2-methoxypyridin
Lieferant: Apollo Scientific
|
Achtung
|
OR11605-1G
OR11605-5G
OR11605-25G
APOSOR11605-1GEA
21.9
EUR
APOSOR11605-1G
APOSOR11605-5G
APOSOR11605-25G
4-Brom-2-methoxypyridin
4-Bromo-2-methoxypyridine
Formel:
C₆H₆BrNO Schmelzpunkt: 25…26 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08277268 CAS-Nummer: 100367-39-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...