5-Bromothiazole-4-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR11641-1G OR11641-250MG
APOSOR11641-1GEA 48 EUR
APOSOR11641-1G APOSOR11641-250MG
5-Bromothiazole-4-carboxylic acid
5-Bromothiazole-4-carboxylic acid
Formel: C₄H₂BrNO₂S
Schmelzpunkt: 182…187 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08275714
CAS-Nummer: 103878-58-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR