5-(2,3-Dichlorphenyl)-4-oxazolcarbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR11671-100MG
OR11671-500MG
APOSOR11671-100MGEA
65.4
EUR
APOSOR11671-100MG
APOSOR11671-500MG
5-(2,3-Dichlorphenyl)-4-oxazolcarbonsäure
5-(2,3-Dichlorophenyl)-4-oxazolecarboxylic acid
Formel:
C₁₀H₅Cl₂NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07376179 CAS-Nummer: 951885-34-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...