5-(2,4-Dichlorphenyl)-4-oxazolcarbonsäure

Lieferant: Apollo Scientific
Achtung

OR11672-1G OR11672-5G OR11672-10G
APOSOR11672-1GEA 134 EUR
APOSOR11672-1G APOSOR11672-5G APOSOR11672-10G
5-(2,4-Dichlorphenyl)-4-oxazolcarbonsäure
5-(2,4-Dichlorophenyl)-4-oxazolecarboxylic acid
Formel: C₁₀H₅Cl₂NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01934490
CAS-Nummer: 255876-52-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR