3,5-Dibrom-4-hydroxybenzoesäure
Lieferant: Apollo Scientific
|
Achtung
|
OR1168-500G
APOSOR1168-500GEA
191
EUR
APOSOR1168-500G
3,5-Dibrom-4-hydroxybenzoesäure
3,5-Dibromo-4-hydroxybenzoic acid
Formel:
C₇H₄Br₂O₃ Molecular Weight: 295.91 g/mol Siedepunkt: 365 Schmelzpunkt: 270…274 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002548 CAS-Nummer: 3337-62-0 EINECS: 222-075-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...