8-Chlor-2-methyl-4-chinazolon
Lieferant: Apollo Scientific
|
Achtung
|
OR11680-1G
OR11680-5G
APOSOR11680-1GEA
464
EUR
APOSOR11680-1G
APOSOR11680-5G
8-Chlor-2-methyl-4-chinazolon
8-Chlor-2-methyl-4-chinazolon
Formel:
C₉H₇ClN₂O Schmelzpunkt: 247…250 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09800989 CAS-Nummer: 19407-54-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...