2-Methoxychinolin-3-boronsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR11687-1G
OR11687-250MG
OR11687-5G
APOSOR11687-1GEA
112
EUR
APOSOR11687-1G
APOSOR11687-250MG
APOSOR11687-5G
2-Methoxychinolin-3-boronsäure
2-Methoxychinolin-3-boronsäure
Formel:
C₁₀H₁₀BNO₃ Molecular Weight: 203,0000 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09800894 CAS-Nummer: 886853-93-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...