2-Thiophen-2-ylpyrrolidine

Lieferant: Apollo Scientific
Achtung

OR11706-1G OR11706-250MG
APOSOR11706-1GEA 393 EUR
APOSOR11706-1G APOSOR11706-250MG
2-Thiophen-2-ylpyrrolidine
2-Thiophen-2-ylpyrrolidine
Formel: C₈H₁₁NS
Molecular Weight: 153,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00052889
CAS-Nummer: 90090-64-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR