5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranoside 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: AICAR, 5-Amino-1-(β-D-ribofuranosyl)-1H-imidazole-4-carboxamide, Acadesine

OR1170T-1G OR1170T-5G OR1170T-250MG
APOSOR1170T-1GEA 109 EUR
APOSOR1170T-1G APOSOR1170T-5G APOSOR1170T-250MG
5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranoside 97%
5-Aminoimidazole-4-carboxamide-1-beta-D-ribofuranoside
Formel: C₉H₁₄N₄O₅
Molecular Weight: 258,23 g/mol
Schmelzpunkt: 214…215 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00869751
CAS-Nummer: 2627-69-2
EINECS: 220-097-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR