Plumbagin
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
5-Hydroxy-2-methylnaphthalene-1,4-dione, 5-Hydroxy-2-methyl-1,4-naphthoquinone
OR1174-1G
OR1174-250MG
APOSOR1174-1GEA
299
EUR
APOSOR1174-1G
APOSOR1174-250MG
Plumbagin
Plumbagin
Formel:
C₁₁H₈O₃ Molecular Weight: 188,18 g/mol Schmelzpunkt: 75…78 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00001682 CAS-Nummer: 481-42-5 EINECS: 207-569-6 UN: 2923 ADR: 8+6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...