4-Hydroxy-3-methoxybenzylalkohol 97+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-Hydroxy-3-methoxyphenyl)methanol, Vanillylalkohol, 2-Hydroxy-5-(hydroxymethyl)anisol
OR1182-500G
APOSOR1182-500GEA
226
EUR
APOSOR1182-500G
4-Hydroxy-3-methoxybenzylalkohol 97+%
4-Hydroxy-3-methoxybenzylalkohol
Formel:
C₈H₁₀O₃ Molecular Weight: 154,17 g/mol Schmelzpunkt: 113…115 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004659 CAS-Nummer: 498-00-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...