3-Chlor-6-(propylamino)pyridazin
Lieferant: Apollo Scientific
Synonyme:
6-Chloro-N-propylpyridazin-3-amine
OR11901-5G
APOSOR11901-5GEA
59.6
EUR
APOSOR11901-5G
3-Chlor-6-(propylamino)pyridazin
3-Chloro-6-(propylamino)pyridazine
Formel:
C₇H₁₀ClN₃ Molecular Weight: 171,6274 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09800940 CAS-Nummer: 951885-19-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...