3-Brombenzanilid

Lieferant: Apollo Scientific

Synonyme: 3-Bromo-N-phenylbenzamide

OR11921-1G OR11921-5G
APOSOR11921-1GEA 21.9 EUR
APOSOR11921-1G APOSOR11921-5G
3-Brombenzanilid
3-Brombenzanilid
Formel: C₁₃H₁₀BrNO
Molecular Weight: 276,1300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00087947
CAS-Nummer: 63710-33-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR