4-Brom-3-methylthioanisol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-2-methyl-4-(methylthio)benzene
OR11944-1G
OR11944-5G
APOSOR11944-1GEA
215
EUR
APOSOR11944-1G
APOSOR11944-5G
4-Brom-3-methylthioanisol 97%
4-Brom-3-methylthioanisol
Formel:
C₈H₉BrS Lagertemperatur: Raumtemperatur |
MDL:
MFCD09801036 CAS-Nummer: 90532-02-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...