1-Boc-isonipecotinsäuremethylester 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl piperidine-4-carboxylate, N-BOC protected, 1-tert-Butyl 4-methyl piperidine-1,4-dicarboxylate
OR11974-100G
OR11974-25G
APOSOR11974-100GEA
40.6
EUR
APOSOR11974-100G
APOSOR11974-25G
1-Boc-isonipecotinsäuremethylester 98%
1-Boc-isonipecotinsäuremethylester
Formel:
C₁₂H₂₁NO₄ Molecular Weight: 243,2994 g/mol Schmelzpunkt: 35 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02183584 CAS-Nummer: 124443-68-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...