2-Amino-6,8-dibromo-3-quinolinecarboxamide

Lieferant: Apollo Scientific

Synonyme: 2-Amino-6,8-dibromoquinoline-3-carboxamide

OR12013-1G OR12013-5G
APOSOR12013-1GEA 270 EUR
APOSOR12013-1G APOSOR12013-5G
2-Amino-6,8-dibromo-3-quinolinecarboxamide
2-Amino-6,8-dibromo-3-quinolinecarboxamide
Formel: C₁₀H₇Br₂N₃O
Molecular Weight: 345 g/mol
Schmelzpunkt: 283…286 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08689708
CAS-Nummer: 937601-68-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR