3-{3-[4-(Benzyloxy)phenoxy]propoxy}benzenecarbaldehyde
Lieferant: Apollo Scientific
OR12029-1G
OR12029-5G
APOSOR12029-1GEA
336
EUR
APOSOR12029-1G
APOSOR12029-5G
3-{3-[4-(Benzyloxy)phenoxy]propoxy}benzenecarbaldehyde
3-{3-[4-(Benzyloxy)phenoxy]propoxy}benzenecarbaldehyde
Formel:
C₂₃H₂₂O₄ Molecular Weight: 362,43 g/mol Schmelzpunkt: 73…75 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08689783 CAS-Nummer: 937602-05-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...