4-{3-[4-(Benzyloxy)phenoxy]propoxy}benzenecarbaldehyde

Lieferant: Apollo Scientific
Achtung

OR12030-1G OR12030-250MG
APOSOR12030-1GEA 415 EUR
APOSOR12030-1G APOSOR12030-250MG
4-{3-[4-(Benzyloxy)phenoxy]propoxy}benzenecarbaldehyde
4-{3-[4-(Benzyloxy)phenoxy]propoxy}benzenecarbaldehyde
Formel: C₂₃H₂₂O₄
Molecular Weight: 362.43 g/mol
Schmelzpunkt: 92…94 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08689784
CAS-Nummer: 937602-07-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR