3'-{3-[4-(Benzyloxy)phenoxy]propoxy}acetophenon
Lieferant: Apollo Scientific
OR12031-1G
OR12031-5G
APOSOR12031-1GEA
336
EUR
APOSOR12031-1G
APOSOR12031-5G
3'-{3-[4-(Benzyloxy)phenoxy]propoxy}acetophenon
3'-{3-[4-(Benzyloxy)phenoxy]propoxy}acetophenon
Formel:
C₂₄H₂₄O₄ Schmelzpunkt: 94…96 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08689785 CAS-Nummer: 937602-09-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...