3-(4-Bromophenyl)-5-methyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Lieferant: Apollo Scientific
OR12047-1G
OR12047-5G
APOSOR12047-1GEA
180
EUR
APOSOR12047-1G
APOSOR12047-5G
3-(4-Bromophenyl)-5-methyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
3-(4-Bromophenyl)-5-methyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Formel:
C₁₅H₁₁BrN₂OS Molecular Weight: 347,24 g/mol Schmelzpunkt: 344…346 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08689700 CAS-Nummer: 937601-63-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...