{1-[(4-Bromophenyl)sulphonyl]-4-piperidinyl}methanol
Lieferant: Apollo Scientific
Synonyme:
1-[(4-Bromophenyl)sulphonyl]-4-(hydroxymethyl)piperidine
OR12048-10G
OR12048-1G
OR12048-5G
APOSOR12048-10GEA
1350
EUR
APOSOR12048-10G
APOSOR12048-1G
APOSOR12048-5G
{1-[(4-Bromophenyl)sulphonyl]-4-piperidinyl}methanol
{1-[(4-Bromophenyl)sulphonyl]-4-piperidinyl}methanol
Formel:
C₁₂H₁₆BrNO₃S Molecular Weight: 334.24 g/mol Schmelzpunkt: 113…115 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08689741 CAS-Nummer: 937012-94-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...