2-Acetyl-6-brompyridin 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR12051-5G
OR12051-1G
OR12051-25G
APOSOR12051-5GEA
24.8
EUR
APOSOR12051-5G
APOSOR12051-1G
APOSOR12051-25G
2-Acetyl-6-brompyridin 98%
2-Acetyl-6-brompyridin
Formel:
C₇H₆BrNO Molecular Weight: 200,03 g/mol Schmelzpunkt: 51…55 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00272200 CAS-Nummer: 49669-13-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...