6-Bromoquinazoline-2,4(1H,3H)-dione

Lieferant: Apollo Scientific
Achtung

OR12053-1G OR12053-25G OR12053-5G
APOSOR12053-1GEA 21.9 EUR
APOSOR12053-1G APOSOR12053-25G APOSOR12053-5G
6-Bromoquinazoline-2,4(1H,3H)-dione
6-Bromoquinazoline-2,4(1H,3H)-dione
Formel: C₈H₅BrN₂O₂
Molecular Weight: 241.04 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00462868
CAS-Nummer: 88145-89-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR