1-[1-(tert-Butoxycarbonyl)-4-piperidinyl]-5-oxo-3-pyrrolidinecarboxylic acid

Lieferant: Apollo Scientific
Achtung

OR12054-1G
APOSOR12054-1GEA 284 EUR
APOSOR12054-1G
1-[1-(tert-Butoxycarbonyl)-4-piperidinyl]-5-oxo-3-pyrrolidinecarboxylic acid
1-[1-(tert-Butoxycarbonyl)-4-piperidinyl]-5-oxo-3-pyrrolidinecarboxylic acid
Formel: C₁₅H₂₄N₂O₅
Molecular Weight: 312.37 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04973994
CAS-Nummer: 937601-51-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR