N-(2-Chloroethyl)-N'-(4-methoxyphenyl)urea

Lieferant: Apollo Scientific

OR12061-1G OR12061-5G
APOSOR12061-1GEA 58 EUR
APOSOR12061-1G APOSOR12061-5G
N-(2-Chloroethyl)-N'-(4-methoxyphenyl)urea
N-(2-Chloroethyl)-N'-(4-methoxyphenyl)urea
Formel: C₁₀H₁₃ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02671069
CAS-Nummer: 65536-40-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR