5-Chloro-2-hydrazinopyridine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Chloro-2-hydrazinylpyridine
OR12065-10G
OR12065-1G
OR12065-5G
APOSOR12065-10GEA
108
EUR
APOSOR12065-10G
APOSOR12065-1G
APOSOR12065-5G
5-Chloro-2-hydrazinopyridine
5-Chloro-2-hydrazinopyridine
Formel:
C₅H₆ClN₃ Molecular Weight: 143,58 g/mol Schmelzpunkt: 127…128 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07786313 CAS-Nummer: 27032-63-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...