6,7-Dichloro-1,4-dihydro-2,3-quinoxalinedione2-oxime

Lieferant: Apollo Scientific

OR12071-10G OR12071-1G OR12071-5G
APOSOR12071-10GEA 894 EUR
APOSOR12071-10G APOSOR12071-1G APOSOR12071-5G
6,7-Dichloro-1,4-dihydro-2,3-quinoxalinedione2-oxime
6,7-Dichloro-1,4-dihydro-2,3-quinoxalinedione2-oxime
Formel: C₈H₅Cl₂N₃O₂
Molecular Weight: 246.05 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08689792
CAS-Nummer: 177944-61-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR