1-(3,4-Dichlorophenyl)tetrahydro-2H-imidazol-2-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(3,4-Dichlorophenyl)imidazolidin-2-one

OR12078-1G OR12078-5G OR12078-500MG
APOSOR12078-1GEA 336 EUR
APOSOR12078-1G APOSOR12078-5G APOSOR12078-500MG
1-(3,4-Dichlorophenyl)tetrahydro-2H-imidazol-2-one
1-(3,4-Dichlorophenyl)tetrahydro-2H-imidazol-2-one
Formel: C₉H₈Cl₂N₂O
Molecular Weight: 231,08 g/mol
Schmelzpunkt: 185…187 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08689734
CAS-Nummer: 52420-29-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR