1,4-Dihydro-2,3-quinoxalinedione-2-oxime 95%

Lieferant: Apollo Scientific

OR12080-10G OR12080-1G OR12080-5G
APOSOR12080-10GEA 596 EUR
APOSOR12080-10G APOSOR12080-1G APOSOR12080-5G
1,4-Dihydro-2,3-quinoxalinedione-2-oxime 95%
1,4-Dihydro-2,3-quinoxalinedione-2-oxime
Formel: C₈H₇N₃O₂
Molecular Weight: 177,16 g/mol
Schmelzpunkt: 262…264 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08689712
CAS-Nummer: 177944-77-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR