2-(Dimethylamino)nicotinonitrile
Lieferant: Apollo Scientific
|
Achtung
|
OR12083-1G
OR12083-250MG
APOSOR12083-1GEA
56.7
EUR
APOSOR12083-1G
APOSOR12083-250MG
2-(Dimethylamino)nicotinonitrile
2-(Dimethylamino)nicotinonitrile
Formel:
C₈H₉N₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08689753 CAS-Nummer: 60138-76-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...