6-(Dimethylamino)nicotinonitrile
Lieferant: Apollo Scientific
|
Achtung
|
OR12084-1G
OR12084-5G
APOSOR12084-1GEA
145
EUR
APOSOR12084-1G
APOSOR12084-5G
6-(Dimethylamino)nicotinonitrile
6-(Dimethylamino)nicotinonitrile
Formel:
C₈H₉N₃ Schmelzpunkt: 66…68 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08272099 CAS-Nummer: 154924-17-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...