3,5-Diaminobenzoesäure
Lieferant: Apollo Scientific
|
Achtung
|
OR1209-250G
OR1209-1KG
OR1209-500G
APOSOR1209-250GEA
34.8
EUR
APOSOR1209-250G
APOSOR1209-1KG
APOSOR1209-500G
3,5-Diaminobenzoesäure
3,5-Diaminobenzoic acid
Formel:
C₇H₈N₂O₂ Molecular Weight: 152,15 g/mol Schmelzpunkt: 235…238 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007807 CAS-Nummer: 535-87-5 EINECS: 208-621-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...