3-Iodo-1H-pyrrolo[2,3-b]pyridine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Iodo-7-azaindole
OR12105-1G
OR12105-25G
APOSOR12105-1GEA
21.9
EUR
APOSOR12105-1G
APOSOR12105-25G
3-Iodo-1H-pyrrolo[2,3-b]pyridine
3-Iodo-1H-pyrrolo[2,3-b]pyridine
Formel:
C₇H₅IN₂ Schmelzpunkt: 190…194 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659023 CAS-Nummer: 23616-57-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...