4-(tert-Butyl)-N-methoxy-N-methylbenzenecarboxamide 95%

Lieferant: Apollo Scientific

Synonyme: 4-(tert-Butyl)-N-methoxy-N-methylbenzamide

OR12107-5G
APOSOR12107-5GEA 806 EUR
APOSOR12107-5G
4-(tert-Butyl)-N-methoxy-N-methylbenzenecarboxamide 95%
4-(tert-Butyl)-N-methoxy-N-methylbenzenecarboxamide
Formel: C₁₃H₁₉NO₂
Molecular Weight: 221,3 g/mol
Schmelzpunkt: 68…70 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06662541
CAS-Nummer: 208188-23-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR